Learn More
Gibco™ Dopaminergic Neuron Maturation Supplement
Dopaminergic Neuron Maturation Supplement is a component of the Gibco™ PSC Dopaminergic Neuron Differentiation Kit that is also offered separately.
Brand: Gibco™ A3147401
This item is not returnable.
View return policy
Description
Dopaminergic Neuron Maturation Supplement is a component of the Gibco™ PSC Dopaminergic Neuron Differentiation Kit that is also offered separately. It enables maturation of excess floor plate progenitor cells to midbrain dopaminergic neurons.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
1 x 10 mL Dopaminergic Neuron Maturation Supplement | |
Neural Cell Culture | |
Cell Culture Supplement | |
6 to 8 | |
Dry Ice |
Stem Cells (Neural) | |
Liquid | |
Sterile | |
10 mL |
For Research Use Only. Not for use in diagnostic procedures.