Learn More
Tocris™ DOPE
Liposome/lipoplex transfection enhancer
Brand:  Tocris™ 7175/25
114.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18768786
  • 146.00 € / 25mg
Explore more special offers
Description
Description
DOPE is a liposome/lipoplex transfection reagent. Enhances liposome and lipoplex transfection efficiency in combination with DA, DOTAP or DC-cholesterol (Cat. No. 7176). Please note: exact ratio for use and transfection efficiency is dependent on cell line used.
Specifications
Specifications
(9 Z)-1,1 '-[(1 R)-1-[[[(2-Aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl] 9-octadecenoic acid ester
25 mg
C41H78NO8P
4004-05-1.
General Laboratory Research Reagent
0.95