Learn More
BRAND™ Dosing Element for BRAND Dispensette S Trace Analysis Dispensers
Replacement dosing element for all models of BRAND Dispensette S Trace Analysis dispensers, 1-10mL. Includes certificate of performance.
Brand:  BRAND™ 708035
386.95 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15613997
  • 580.00 € / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement dosing element for BRAND Dispensette S Trace Analysis 1-10mL
  • Fits instruments with tantalum or platinum-iridium spring
  • Includes certificate of performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL
Dispensette™ S Trace Analysis
Quality Certificate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Dosing Element for BRAND Dispensette S Trace Analysis Dispensers > Dosing element for BRAND Dispensetts S Trace Analysis (all models)