Learn More
Miele Professional™ Dosing System
Hose dispenser pump, with level indicator for the automatic dispensing of liquid cleaning agents
Brand:  Miele Professional™ 9968660
 View more versions of this product
Product Code. 15487714
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For Miele glass laboratory washer PG 8504
  • Dispenser module for liquid detergent
  • 20cm siphon for canister
  • 'Container empty' indicator
Specifications
Specifications
Gray
5 L
200 mm
200/230 V