missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL DOSIPET B
Dosipet B is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.
Brand: KARTELL 8301000
This item is not returnable.
View return policy
Description
- For universal use; ideal for use with reagent of average aggressiveness.
- It can be used with every water solution type (except those containing hydrofluoric acid), marked by a high degree of convenience and reliability. It is equipped with a 3.3 borosilicate glass cylinder and PTFE piston.
- Precise and quick volume adjustment using the analogue slider
- Precise graduated scale to ensure reproducibility of the dispensed volumes
- No loss of reagent when eliminating air or filling
- Dispensing without bubbles forming
- The piston completely transfers the liquid with every movement
- Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
- Autoclavable at +121°C for 20 minutes
Specifications
± 0.06 mL | |
Autoclavable | |
White | |
Borosilicate Glass, PTFE | |
Analog | |
0.34 oz. |
Dosipet B | |
None | |
1 | |
10 mL | |
0.20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction