missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL DOSIPET S
Dosipet S is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.
497.00 € - 790.00 €
Specifications
Product Type | Dosipet S |
---|---|
Autoclavable | Autoclavable |
Color | White |
Channels | 1 |
Material | Borosilicate Glass, PTFE |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10020263
|
KARTELL
8307000 |
10 mL |
497.00 €
Each |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
10253143
|
KARTELL
8307100 |
25 mL |
700.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10059782
|
KARTELL
8307200 |
50 mL |
790.00 €
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
Description
- Useful for dosing concentrated acids, polar and no-polar solvents.
- High quality components and assembly make this series the ideal choice for measuring out organic polar and non- polar solvents, including halogenated solvents, and concentrated mineral acids, including particularly volatile acids such as nitric and trifluoroacetic acids.
- In addition, they can be used with peroxides, hydrochloric acid and sulphuric acid.
- It comes with a borosilicate glass piston.
- The reagents come into contact only with parts made of PTFE
- Precise and quick volume adjustment using the analogue slider
- Precise graduated scale to ensure reproducibility of the dispensed volumes
- No loss of reagent when eliminating air or filling
- Dispensing without bubbles forming
- The piston completely transfers the liquid with every movement
- Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
- Autoclavable at +121°C for 20 minutes
Specifications
Dosipet S | |
White | |
Borosilicate Glass, PTFE |
Autoclavable | |
1 | |
Analog |