missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Double Bagged - USP Class VI Liquid Funnels
BIOPHARMA

Our premium range of Liquid Funnels are made from high grade medical USP Class VI HDPE. It is ideal for demanding applications such as vaccine manufacture, GMP, and cleanroom manufacturing.

85.15 € - 137.00 €

Specifications

Material HDPE
Sterility Sterile
View More Specs

Products 2
Product Code Brand Diameter (Metric)Top Price Quantity & Availability  
Product Code Brand Diameter (Metric)Top Price Quantity & Availability  
17594706
BIOPHARMA
View Documents
Sampling Systems™
9370H-50S
50 mm
137.00 €
Pack of 50
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
17584706
BIOPHARMA
View Documents
Sampling Systems™
9370H-100S
100 mm
85.15 €
Pack of 20
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
Description

Description

  • The SteriWare™ Ultra range of liquid funnels are available in both 50 mm and 100 mm diameter.
  • This premium funnel range is double bagged.
  • A box liner is also used giving a third layer of protection.
  • It is ideal for demanding applications such as vaccine manufacture, GMP and cleanroom manufacturing.
  • The funnels all have an ergonomic lug on the side so handling them cleanly is easy.
  • The funnels are designed to be used straight from the bag.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The funnels are sterilised by gamma irradiation.
  • USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • Cleanroom Manufactured - The funnels are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each funnel is 100% inspected before packing.
Specifications

Specifications

HDPE
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.