Learn More
Brand™ Double Ruled Without Clips Counting Chamber Fuchs-Rosenthal
Brand:  Brand™ 719805
55.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 15623997
  • 61.50 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Counting Chamber Fuchs-Rosenthal
16 mm2
CE-marked according to IVD 98/79 EG.
0.2 mm