Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Double-sided crosshead
Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on both sides of the stirrer for greater turbulence.
Brand:  Fisherbrand™ 001.1160
 View more versions of this product
Product Code. 10211812
  • 10.15 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Double-sided crossheads provider greater turbulence during mixing
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
PTFE
2/Pk.
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
15mm
60 mm
PTFE Stirrer Bars Crosshead