missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin hydrochloride, Tocris Bioscience™

Product Code. 18732474
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antitumor antibiotic agent. Inhibits DNA topoisomerase II

Doxorubicin hydrochloride is a 14-hydroxylated version of Daunorubicin (Cat. No. 1467) that is naturally fluorescent. Doxorubicin is an antitumor antibiotic agent that inhibits DNA topoisomerase II. It is a DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Doxorubicin reduces intracellular tau levels. Doxorubicin also promotes formation of free radicals for the disruption of membrane lipids and DNA strands. Doxorubicin fluorescence is quenched upon intercalation into the DNA; while binding to histones or partitioning into the phospholipid phase of PEG-phospholipid micelles or hydrophobic cores of polymeric micelles, increases Doxorubicin fluorescence. Doxorubicin fluorescence enables the monitoring of the localization of the drug within lipid bilayers and liposomal delivery systems and interaction of the drug with DNA and other macromolecules, as well as drug efflux pump activities.

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Quantity 10 mg
Formula Weight 579.99
Percent Purity >98%
Chemical Name or Material Doxorubicin hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'