missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline (hyclate), MedChemExpress

Product Code. p-200098729
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16461438

Brand: Medchem Express HYN0565B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity.

Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity.

Specifications

Chemical Name or Material Doxycycline (hyclate)
CAS 24390-14-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H24N2O8.1/2C2H6O.ClH.1/2H2O
Quantity 5 g
Synonym Doxycycline hydrochloride hemiethanolate hemihydrate WC2031
Solubility Information DMSO : 240 mg/mL (467.89 mM; Need ultrasonic) ∣H2O : 125 mg/mL (243.69 mM; Need ultrasonic)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])[C@@H](O)[C@]3([H])[C@@H](C)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N.Cl.[0.5H2O].[0.5C2H6O]
Molecular Weight (g/mol) 512.94
Formula Weight 512.94
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.