missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™

Broad spectrum antibiotic

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
View More Specs

Description

Description

Broad spectrum antibiotic

Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.
Specifications

Chemical Identifiers

10592-13-9
480.90
VLUQVUWDECWBTL-UQVCFKGQSA-N
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
C22H25ClN2O8
MFCD03427564
54706018
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Doxycycline Hydrochloride
Inclusive between 300 at 349nm
210°C
C22H25ClN2O8
Inclusive between 800μg/mg
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
480.90
480.9
Inclusive between 1.4%
Doxycycline Hydrochloride
Powder/Solid
10592-13-9
95%
MFCD03427564
VLUQVUWDECWBTL-UQVCFKGQSA-N
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
54706018
Inclusive between 105°
Yellow
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not induce vomiting. Call a physician or Poison Control Center immediately. . .

NFPA

Health:2
Flammability:0
Instability:0

Recommended Storage : 0° to 5°C, light sensitive, desiccate.

Product Identifier
  • Doxycycline Hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.