missing translation for 'onlineSavingsMsg'
Learn More

Doxylamine succinate, 98%, Thermo Scientific Chemicals

Product Code. 13545040
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-10-7
Molecular Formula C21H28N2O5
Molecular Weight (g/mol) 388.46
MDL Number MFCD00056168
InChI Key KBAUFVUYFNWQFM-UHFFFAOYNA-N
Synonym doxylamine succinate, decapryn, evigoa d, a-h injection, decapryn succinate, unisom, alsadorm, gittalun, mereprine
PubChem CID 11224
ChEBI CHEBI:82461
IUPAC Name butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
SMILES OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 g
Formula Weight 388.46
Chemical Name or Material Doxylamine succinate
compliance-icons
Product Identifier
  • Doxylamine succinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'