missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, no calcium, no magnesium
GREENER_CHOICE

DPBS, no calcium, no magnesium

11.06 € - 734.00 €

Specifications

Content And Storage 15°to 30°C
Dilution 1 X
Product Type DPBS
Form Liquid
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11580476
GREENER_CHOICE
View Documents
Gibco™
14190086
100 mL
11.06 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11590476
GREENER_CHOICE
View Documents
Gibco™
14190094
500mL
24.84 €
500mL
Please to purchase this item. Need a web account? Register with us today!
12047539
GREENER_CHOICE
View Documents
Gibco™
14190185
20 x 100mL
204.00 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
12037539
GREENER_CHOICE
View Documents
Gibco™
14190169
10 x 500 mL
217.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
12008489
View Documents
Gibco™
14190240
10 L
734.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


Universal Bag Packaging
Gibco™ universal bags offer flexible packaging that allows you to integrate a single-use, disposable large volume media container with a wide variety of automated cell culture instruments, disposable bioreactor systems, and sterile connection processes, minimizing the need for costly customization.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications

Specifications

15°to 30°C
1 X
DPBS
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 36 Month(s)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ DPBS, no calcium, no magnesium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.