Learn More
Lovibond™ DPD No.1 Tablet
A high quality comparator instrument for visually matching the intensity of sample color with Lovibond™ glass filters
Brand:  Lovibond™ 511051BT
Product Code. 13151681
  • 57.10 € / 250 tablets
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
A high quality comparator instrument for visually matching the intensity of sample color with Lovibond™ glass filters
Lovibond™ Comparator 2000+
A high quality comparator instrument for visually matching the intensity of sample color with Lovibond™ glass filters. It includes an integrated prism, which brings the sample and comparison fields together, allowing enhanced color discrimination and matching. The prism is ultrasonically sealed to prevent contamination of the optics. It includes the facility to compensate for water samples that are turbid or tinted.
Lovibond™ Daylight 2000 unit
A standardised bench top light source that guarantees constant and uniform lighting conditions for color matching with the Lovibond™ Comparator 2000+ instruments (comparator not included).
Lovibond™ Water Test Discs and Reagents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. Some of the more popular discs are listed here but other ranges and tests can be supplied on request (for example chlorine dioxide, copper, fluoride, hydrazine, hydrogen peroxide, iodine, manganese, molybdate, nitrite, oxygen, ozone, phosphate, QAC, silica, sulfide, zinc).
What you need
  • Comparator instrument (appropriate Nessleriser)
  • Lovibond™ disc, specific to the test
  • Reagents, specific to the test
  • Sample container of the required path length
  • Lighting unit (optional) to ensure constant viewing conditions
  • Simple and flexible system giving reliable results in both laboratory and field testing
  • Compact, easily portable and robust for field use
  • Reagents mostly available in convenient tablet form
  • Includes stable colored glass filters which can be certified under a ISO 9001 QA procedure
Specifications
Specifications
250/Pk.
250/Pk.