Learn More
DPPC, MedChemExpress
MedChemExpress DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.
41.85 € - 67.50 €
Specifications
Chemical Name or Material | DPPC |
---|---|
CAS | 63-89-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C40H80NO8P |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484298
|
Medchem Express
HY-109506 |
10 mg |
41.85 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480627
|
Medchem Express
HY-109506 |
50 mg |
67.50 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.Specifications
DPPC | |
White | |
C40H80NO8P | |
Ethanol : 25 mg/mL (34.06 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (6.81 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
734.04 | |
98.0% | |
Solid |
63-89-8 | |
Research | |
129Y83 | |
CCCCCCCCCCCCCCCC(O[C@H](COC(CCCCCCCCCCCCCCC)=O)COP(OCC[N+](C)(C)C)([O-])=O)=O | |
734.04 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.