missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ XXS Replacement Ammonia DragerSensor™

For Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors.

Brand:  Dräger™ 6810888

Product Code. 15520713

  • 540.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Measuring Range: 0 to 300ppm
  • Resolution: 1ppm

Services

One year

Specifications

Specifications

0 to 300ppm
Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors
Accurate detection of Ammonia
Ammonia
Ammonia Replacement Sensor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.