missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Carbon Monoxide XS Replacement Sensor

For use with X-am 7000 Multi-Gas Monitor

Brand:  Dräger™ 6809105

Product Code. 11342090

  • 483.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Detection Range: 0-2000ppm
  • Detects: CO

Services

3 Year

Specifications

Specifications

Carbon Monoxide (9XS)
0 to 2000
1
-40° to +48.8°C
10 to 90%
700 to 1300mbar
-40°F to +120°F
3 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers