Learn More
Dräger™ CMS Chips: Ethanol
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406370
Product Code. 10287467
  • 87.30 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 25mg/L
60 to 340 sec.
10°C to 30°C
Ethanol CMS Chip
100 to 2500
50°F to 86°F
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • GAS SAMPLING CASSETTE DRAEGER CMS CHIP ETHANOL 10
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid