Learn More
Dräger™ CMS Chips: Styrene
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406560
Product Code. 12314649
  • 94.75 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 30mg/L
100 to 550 sec.
5° to 40°C
Styrene CMS Chip
2 to 40
41°F to 104°F