Learn More
Dräger™ CMS Chips: Trichloroethylene
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406320
Product Code. 10056001
  • 82.45 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
40 to 330 sec.
0°C to 40°C
Trichloroethylen CMS Chip
5 to 100
32°F to 104°F