Learn More
Dräger™ CMS Chips: Methanol
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406380
Product Code. 15442273
  • 106.00 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 25mg/L
200 to 600 sec.
15°C to 30°C
Methanol CMS Chip
20 to 500
59°F to 104°F