Learn More
Dräger™ CMS Chips: Xylene
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406260
Product Code. 10682333
  • 130.00 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
75 to 500 sec.
0°C to 40°C
o-Xylene CMS Chip
10 to 300
32°F to 104°F