Learn More
Dräger™ CMS Chips: MTBE
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406530
Product Code. 12344629
  • 91.05 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
90 to 450 sec.
10°C to 30°C
MTBE CMS Chip
10 to 200
50°F to 86°F