Learn More
Dräger™ CMS Chips: Petroleum Hydrocarbons
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406200
 View more versions of this product
Product Code. 10446301
  • 114.00 € / Each
Explore more special offers
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
150 to 330 sec.
0°C to 40°C
Petroleum Hydrocarbons CMS Chip
20 to 500
32°F to 104°F