Learn More
Dräger™ Connecting cradle for Pac™ 1000, 3500, 5500, and 7000 Series Gas Detectors
Accessory for Pac series gas detectors
Brand:  Dräger™ 8318587
Product Code. 15120629
  • 260.00 € / Each
Explore more special offers
Includes: USB cable
Description
Description
  • Allows connection of any Pac detector to PC
Specifications
Specifications
10 x 8 x 1 in. (304.8 x 20.32 x 2.5cm)
Connecting Cradle
2.5 cm
304.8 cm
20.32 cm
Pac 1000, 3500, 5500, and 7000 Series Gas Detectors
1 in.
10 ft.
8 in.