missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Hydrogen Sulfide (H2S) XXS Replacement Sensor

XXS Replacement Sensor, for use with Pac 7000, X-am 2000, X-am 5000 multi-gas detectors.

Brand:  Dräger™ 6810883

Product Code. 15513975

  • 309.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Measuring range: 0 to 100 ppm
  • Resolution: 1 ppm
  • Warranty: 2 years
  • Hydrogen Sulfide (H2S)
Specifications

Specifications

Hydrogen Sulfide
Pac 7000, X-am 5000, X-am 5600 Multigas Detectors
Replacement Sensor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.