Learn More
Dräger™ Individual Haz Mat Simultaneous Test Sets
For simultaneous testing of five different gases
82.80 €
Specifications
For Use With (Application) | For simultaneous testing of five different gases |
---|
Alert:
No other type of tube should be substituted with these kits.
Requires: Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
12702517
|
Dräger™
8101735 |
Simultaneous Test Set I | Tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes |
82.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10385704
|
Dräger™
8101736 |
Simultaneous Test Set II | Tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene |
82.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10345754
|
Dräger™
8101770 |
Simultaneous Test Set III | Tests for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones |
82.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Test Set I. Includes detector tubes for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes.
Test Set II. Includes detector tubes for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene.
Test Set III. Includes detector tubes for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones.
Order Info
Packs of 10 test sets.
Specifications
For simultaneous testing of five different gases |