Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Brand:  Dräger™ CH29101
 View more versions of this product
Product Code. 10276821
  • 98.95 € / Pack of 10
Explore more special offers
Specifications
Hydrogen Sulfide 100/a
100 to 2000ppm
10
32°F to 104°F
3 to 40mg/L
1
30 sec.
0°C to 40°C