Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Brand:  Dräger™ CH29801
 View more versions of this product
Product Code. 10378421
  • 108.00 € / Pack of 10
Explore more special offers
Specifications
Hydrogen Sulfide 5/b
5 to 60ppm
10
32°F to 140°F
Less than40mg/L
10
4 min.
0°C to 60°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • X10 GAS DETECTOR TUBES DRAEGER CH29801 HYDROGEN
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen sulphide