missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dräger™ Short-Term Detector Tubes: Oil Mist
Gas detection tubes for oil mist.
Brand: Dräger™ 6733031
This item is not returnable.
View return policy
Description
Gas detection tubes for oil mist.
6733031:
- Indicates only mineral and synthetic oil aerosols (mist)
- Standard measuring range: 1 to 10mg/m3
- No.of Strokes n: 100
- Time for measurement: approx. 25 min.
- Color change: White to brown
6728371:
- Indicates the total concentration of mineral and synthetic oil aerosols (mist) and oil vapors
- Standard measuring range: 25 to 100ug
- Color change: White to beige or yellow
Order Info
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Safety Equipment Institute (SEI) certified | |
Less than20mg/L | |
100 | |
25 min. | |
10°C to 30°C |
Oil Mist 1/a | |
1 to 10mg/m3 | |
10 | |
50°F to 86°F |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
- X10 GAS DETECTOR TUBE DRAEGER 6733031 OIL MIST
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.