Learn More
Dräger™ Short-Term Detector Tubes: Water Vapor
Detector tubes
93.20 € - 220.00 €
Specifications
Components Measured Water Vapor
Quantity 10
View More Specs
Products 5
Product Code Brand Components Measured Tube Type Measurement Range Response Time Price Quantity & Availability  
Product Code Brand Components Measured Tube Type Measurement Range Response Time Price Quantity & Availability  
10318562
View Documents
Dräger™
6728531
Water Vapor Water vapor 5/a-p 5 to 200 25 min.
220.00 €
Pack of 10
15242770
View Documents
Dräger™
8103031
Water Vapor Water vapor 3/a 3 to 60 Not Applicable
116.00 €
Pack of 3
10729314
View Documents
Dräger™
8101781
Water Vapor Water vapor 1/b 20 to 40mg/L 20/40 sec.
160.00 €
Pack of 10
10296602
View Documents
Dräger™
8103061
Water Vapor Water vapor 20/a-P 20 to 1500 10 to 5 min.
198.00 €
Each
10062541
View Documents
Dräger™
CH23401
Water Vapor Water vapor 0.1 1 to 40 2 min.
93.20 €
Each
Description
Description
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ Customer Service for more information.
Specifications
Specifications
Water Vapor
10
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Dräger™ Short-Term Detector Tubes: Water Vapor