Learn More
Dräger™ XS-2 Sensors for MiniWarn™ Multi-Gas Monitors
For Detecting or monitoring oxygen gas
Brand:  Dräger™ 6810375
Product Code. 11322080
  • 337.00 € / Each
Explore more special offers
Description
Description
  • Measuring range of 0-25% volume
  • 0.1% volume resolution
  • Compatible with Pac III, MiniWarn and Multiwarn II monitors
  • Services
    2 years
Specifications
Specifications
Oxygen
0 to 25%
0.1
-40° to +50°C
10 to 95%
700 to 1300mbar
-40°F to +122°F
2 Years