Learn More
Draeger Safety DrögerSensor™ XXS CO LC
Draeger Safety DrögerSensor™ XXS CO LC detects CO at 1 ppm resolution for use with X-am™ 5600 series multi-gas monitors.
Brand:  Draeger Safety 6813210
Product Code. 15600899
  • 309.00 € / Each
Explore more special offers
Description
Description
  • 0 - 2000 ppm gas detector sensor
  • Detects CO at 1 ppm resolution
  • For use with X-am™ 5600 series multi-gas monitors
Specifications
Specifications
CSA Certified, MED Approved
X-am 5600 Series Multigas Monitors
1 ppm
Carbon Monoxide
0 to 2000 ppm
-20°C to +50°C