Learn More
Hirschmann™ Automatic Burettes, Acc. to Dr. Schilling
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Automatic Burettes, Acc. to Dr. Schilling are available in a range of capacities to fit your needs.
Brand:  Hirschmann™ 3600164
 View more versions of this product
Product Code. 10569222
  • 90.45 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With Schellbach stripe
  • Blue graduation
  • Fast flow burette
Specifications
Specifications
15 mL
1 mL
0.045
Batch, single certificate and a DKD Certificate (former DKD Calibration)
Glass
LDPE Bottle
Titration Apparatus