missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Ammonia

Detector tubes

79.00 € - 164.00 €

Specifications

Description Detector Tubes
Components Measured Ammonia
Quantity 10
View More Specs

Products 6
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
10060042
View Documents
Dräger™
8101711
Ammonia Ammonia 0.25/a 0.25 to 3 Less than20mg/L
163.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
10110971
View Documents
Dräger™
8101941
Ammonia Ammonia 5/b 5 to 100 Less than20mg/L
134.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
10299941
View Documents
Dräger™
6733231
Ammonia Ammonia 2/a 2 to 30 Less than20mg/L
139.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15220578
View Documents
Dräger™
6733231
Ammonia Ammonia 2/a 2 to 30 Less than20mg/L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10695431
View Documents
Dräger™
CH20501
Ammonia Ammonia 5/a 5 to 700 Less than20mg/L
164.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15132323
View Documents
Dräger™
CH31901
Ammonia Ammonia 0.5%/a 0.5 to 10% 3 to 12mg/L
120.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Specifications

Detector Tubes
Ammonia
10
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.