missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRAQ5™ Fluorescent Probe Solution (5 mM)

Stain live or fixed cells with this far-red DNA stain.

Brand:  Thermo Scientific™ 62254

285.95 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.



 View more versions of this product

Product Code. 11820154

  • 308.00 € / 50µL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Stain live or fixed cells with Thermo Scientific™ DRAQ5 Fluorescent Probe, a far-red DNA stain.

Fluorescence cell-based assays, such as flow cytometry, in-cell ELISA, fluorescence microscopy and high-content imaging require a fluorescent label to identify individual cells. When using multiple fluorescent probes to detect different cellular targets or activities, each probe must have a fluorescent spectrum different than the other probes. The blue-fluorescent DNA-binding probes, Hoechst and DAPI are frequently used; however, these probes cannot be used when UV illumination is unavailable or other blue-emitting fluorescent probes are used. Therefore, nuclear probes that emit in a color other than blue is useful for cell identification and counting, and for determining nuclear morphology and DNA content.
  • Due to its far-red excitation and emission, the DRAQ5 Fluorescent Probe can be multiplexed with many other fluorophores and is ideal for cells expressing green fluorescent protein (GFP) fusion proteins
  • Compatible with many existing protocols across a wide range of instrumentation platforms
  • Emits in the far-red region, is lipophilic and crosses cell and nuclear membranes in live and fixed cells and tissues for rapid DNA staining
  • Stain is water-soluble, supplied ready-to-use and does not require RNase, cell lysis, or a washing step, making it compatible with automation
  • Because DNA staining is stoichiometric, the DRAQ5 Fluorescent Probe can be used for DNA content analysis in cell proliferation studies
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell-permeant
Store at 4°C.
Fluorescence
Fluorescence Microscope, High Content Analysis Instrument
Cell-Permeant
647 nm
Pierce™
50 μL
Nucleus, Cytosol, Cytoplasm
Red
Cellomics DRAQ5 Fluorescent Probe
Cell Imaging
Fluorescent Probe Solution
681 nm
Liquid
> 99%
Soluble at working concentrations in water and typical buffers
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DRAQ5 50 UL (5MM)

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : 1, 5-bis{¢2-(dimethylamino)ethyl!amino }-4, 8-dihydroxyanthracene-9, 1 0-dione( 0.1-1 ), 1, 5-bis{

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.