Learn More
Thermo Scientific™ Drawer Dividers
Maximize storage inside blood bank refrigerators and plasma freezers with these optional drawer dividers.
232.16 €
Products 1
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11904286
View Documents Promotion Details
Thermo Scientific™
7027
4.9 and 11.5 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
232.16 €
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
Description
Description
Maximize storage inside blood bank refrigerators and plasma freezers with these optional Thermo Scientific™ Drawer Dividers.
Stainless-steel drawer dividers help manage inventory of bags and boxes:
  • Four column dividers for all models except 29.2 cu. ft. (826L) models
  • Five column dividers for 29.2 cu. ft. (826L) models
  • Warranty varies by country, please contact us for details
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.