missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DREADD agonist 21
Potent hM3Dq and hM4Di DREADD agonist; blood brain barrier penetrant
286.00 € - 1070.00 €
Specifications
Chemical Name or Material | 11-(1-Piperazinyl)-5H-dibenzo[b,e][1,4]diazepine |
---|---|
CAS | 56296-18-5 |
Target | DREADD Activators |
Molecular Formula | C17H18N4 |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18025422
|
Tocris Bioscience™
5548/10 |
10 mg |
286.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18065292
|
Tocris Bioscience™
5548/50 |
50 mg |
1070.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DREADD agonist 21 is a potent muscarinic DREADD agonist (pEC50 values are 8.91, 8.48 and 7.77 for hM1Dq, hM3Dq and hM4Di in vitro, respectively). Exhibits > 10-fold higher affinity for hM1Dq and hM4Di compared to wild type receptors. Also displays little to no activity at wild type M3 receptors. Activates neurons expressing hM3Dq DREADDS and inhibits activity in neurons expressing hM4Di in vivo. Displays excellent brain permeability. Water soluble salt also available, DREADD agonist 21 hydrochloride (Cat. No. 6422).Specifications
11-(1-Piperazinyl)-5H-dibenzo[b,e][1,4]diazepine | |
DREADD Activators | |
0.98 |
56296-18-5 | |
C17H18N4 |
Safety and Handling
Recommended Storage : Store at Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title