missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Buffers (10X)

Thermo Scientific 10X DreamTaq Buffer is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase.

Brand:  Thermo Scientific™ B65

16.52 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11856863

  • 25.41 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific DreamTaq Buffer (10X), available in colorless and green formats, is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase. It contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2

In addition, 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes. The density reagent allows direct loading of PCR products on a gel. The blue dye (migrates with 3–5 kb DNA fragments in 1% agarose gel) and the yellow dye (migrates faster than 10 bp DNA fragments in 1% agarose gel) are included for monitoring electrophoresis progress. The dyes have excitation peaks at 424 nm and 615 nm.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless DreamTaq Buffer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 10X DreamTaq Buffer (with 20 mM MgCl2), 4 x 1.25 mL

Store in a non-frost-free freezer at -20°C.
4 x 1.25 mL
Hot-start PCR, Standard PCR
1.25 mL
DreamTaq Hot Start DNA Polymerase, DreamTaq DNA Polymerase
Wet or Dry Ice
10X
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.