missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq PCR Master Mix (2X)

Get higher sensitivity, longer PCR products and higher yields in all standard PCR applications with this enhanced Taq DNA polymerase.

Brand:  Thermo Scientific™ K1071

85.15 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 11896833

  • 131.00 € / 200 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq DNA Polymerase uses the same reaction set-up and cycling conditions as conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required. It is supplied with optimized DreamTaq buffer, which includes 20 mM MgCl2.

DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR set up. The master mix retains all features of DreamTaq DNA Polymerase. It is capable of robust amplification of up to 6 kb from genomic DNA and up to 20 kb from viral DNA.

Highlights

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 4 x 1.25 mL DreamTaq PCR Master Mix (2X), which includes DreamTaq DNA Polymerase, 2X DreamTaq buffer, dNTPs, and 4 mM MgCl2
• 4 x 1.25 mL Nuclease-free water

Store at -20°C.
DreamTaq DNA Polymerase
1 X
3'-A
Low
Standard
No
SuperMix or Master Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.