missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase

Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

Brand:  Thermo Scientific™ EP1714

2319.00 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.


 View more versions of this product

Product Code. 15659374

  • 3865.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Green buffer that includes density reagent and two tracking dyes for convenient direct loading of PCR products on a gel. The buffer contains magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available without pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.

DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases.

DreamTaq Hot Start Green DNA Polymerase is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths—routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction set-up
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Direct loading of PCR products on a gel with DreamTaq Green buffer

Applications

  • Routine PCR
  • Colony PCR
  • Genotyping
  • RT-PCR
  • Generation of PCR products for TA cloning

Recommended Storage

Store at -20C.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• DreamTaq Hot Start DNA Polymerase (5 U/μL), 20 x 100 μL
• 10X DreamTaq Green Buffer (with 20 mM MgCl2), 40 x 1.25 mL

Store at -20°C.
DreamTaq DNA Polymerase
4 x 2500 Units
Hot-start PCR
Built-In Hot Start
Separate Components
Low
Standard
Dry Ice
5 U/μL
3'-A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers