missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DreamTaq DreamTaq™ Hot Start Green PCR Master Mix
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
Brand: DreamTaq K9021
Includes: DreamTaq Hot Start Green PCR Master Mix, 2X; water, nuclease-free
Description
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.Better reaction outcomes:
- Higher yield of target amplicons from low template amounts.
- Increased reaction specificity due to reliable hot-start technology.
- Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
- Room temperature reaction set-up.
- Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
- Simplified workflow and minimized pipetting steps with 2X master mix format.
Fidelity (vs. Taq): 1X
Recommended Storage
Store at -20C.
Specifications
Contains: • 4 x 1.25 mL DreamTaq Hot Start Green PCR Master Mix (2X) • 4 x 1.25 mL Nuclease-free water Store at -5 to -30°C. |
|
DreamTaq DNA Polymerase | |
Wet or Dry Ice | |
3'-A |
Low | |
Standard | |
Built-In Hot Start | |
SuperMix or Master Mix |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction