missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start PCR Master Mixes

Thermo Scientific DreamTaq Hot Start PCR Master Mix is a ready-to-use 2X reaction mix that includes DreamTaq Hot Start DNA Polymerase, DreamTaq buffer, magnesium, and dNTPs.

131.00 € - 760.00 €

Specifications

GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
View More Specs

Products 7
Product Code Brand Color No. of Reactions Price Quantity & Availability  
Product Code Brand Color No. of Reactions Price Quantity & Availability  
15669374
View Documents
Thermo Scientific™
K9011
Colorless 200 Reactions
177.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15717536
View Documents
Thermo Scientific™
K9011
Colorless 200 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15686154
View Documents
Thermo Scientific™
K9012
Colorless 1000 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15679374
View Documents
Thermo Scientific™
K9012
Colorless 1000 Reactions
760.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15737536
View Documents
Thermo Scientific™
K9021
Green 200 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15747536
View Documents
Thermo Scientific™
K9022
Green 1000 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15606164
View Documents
Thermo Scientific™
K9022
Green 1000 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Thermo Scientific DreamTaq Hot Start PCR Master Mix, available in colorless and green formats, is a ready-to-use 2X reaction mix that includes DreamTaq Hot Start DNA Polymerase, DreamTaq Buffer, magnesium, and dNTPs.

The master mix allows easy reaction setup and fewer pipetting steps, with only primers, template, and water needing to be added. The DreamTaq Hot Start Green PCR Master Mix includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases.

DreamTaq Hot Start PCR Master Mix is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Simplified workflow and minimized pipetting steps with 2X master mix format
    • Direct loading of PCR products on a gel with green format

Applications

  • Generation of PCR products for TA cloning
  • Routine PCR
  • Colony PCR
  • Genotyping
  • RT-PCR
Specifications

Specifications

Low
Standard
Hot-start PCR
Built-In Hot Start
SuperMix or Master Mix
DreamTaq DNA Polymerase
Dry Ice
2X
3'-A
Videos
SDS
Documents

Documents

Product Certifications