missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq PCR Master Mixes (2X)

Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.

Brand:  Thermo Scientific™ K1072

 View more versions of this product

Product Code. 15797516

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific DreamTaq PCR Master Mix (2X), available in colorless and green formats, is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR setup.

The master mix retains all features of DreamTaq DNA Polymerase and is capable of robust amplification up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.

DreamTaq Green PCR Master Mix (2X) additionally includes 2X DreamTaq Green Buffer with a density reagent and two dyes for monitoring electrophoresis progress.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• DreamTaq PCR Master Mix (2X), 20 x 1.25 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -20°C.
DreamTaq DNA Polymerase
20 x 1.25 mL
Standard PCR
1X
3'-A
Colorless
Low
Standard
Dry Ice
5 U/μL
No
SuperMix or Master Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2821351 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1072
Lot Number2821359 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1072
Lot Number2791462 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1072
Lot Number2794307 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1072
Lot Number2870724 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberK1072
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DreamTaq PCR Master Mixes (2X) > Color: Colorless, Quantity: 20 x 1.25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.