Learn More
Lenz Laborglasintrumente™ Drechsel Gas Washing Bottles
Gas washing bottles acc. to Drechsel with filter disc
Brand:  Lenz Laborglasintrumente™ 05510270
 View more versions of this product
Product Code. 10622092
  • 92.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12596
  • Complete with head (with filter disc)
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Specifications
1000 mL
Olive Connections
DURAN™ tubing head, borosilicate glass 3.3 bottle
2