missing translation for 'onlineSavingsMsg'
Learn More
Drierite™, with indicator, 4 mesh, Thermo Scientific Chemicals Product Code.: 10742071

Drierite™, with indicator, 4 mesh, Thermo Scientific Chemicals

Product Code. 10742071
500 g, Glass Bottle
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10742071

Brand: Thermo Scientific Acros 219085000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9, 7646-79-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Melting Point 1450.0°C
Physical Form Irregular Granules or Chunks
Material Calcium Sulfate with Cobalt Chloride
Quantity 500 g
Fieser 01,107
Merck Index 15, 1708
Packaging Glass Bottle
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
IUPAC Name calcium;sulfate
Formula Weight 136.13
Chemical Name or Material Drierite™, 4 mesh
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467668 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number219085000,219080100,219080020
Lot NumberA0467665 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number219085000,219080100,219080020
Lot NumberA0467667 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number219085000,219080100,219080020
Lot NumberA0467666 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number219085000,219080100,219080020
Lot NumberA0464752 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number219085000,219080100,219080020
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Calcium sulfate, Cobalt(II) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Drierite™, with indicator, 4 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.