Learn More
Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
Drierite, CaO4S, CAS Number-7778-18-9, 7646-79-9, 5kg, 39.09%, 0.68%, CHEBI:31346, 3.25%, Blue, 01,107, 136.13, GHS Signal Word: Danger
116.00 € - 694.00 €
Chemical Identifiers
CAS | 7778-18-9, 7646-79-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
PubChem CID | 24497 |
ChEBI | CHEBI:31346 |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10276750
|
Thermo Scientific Acros
|
500 g | Glass Bottle |
116.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10082880
|
Thermo Scientific Acros
219090020 |
2 kg | Glass Bottle |
332.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10722934
|
Thermo Scientific Acros
219090050 |
5 kg | Plastic Drum |
694.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, IndicatorChemical Identifiers
7778-18-9, 7646-79-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
CHEBI:31346 |
CaO4S | |
MFCD00010912 | |
24497 | |
[Ca++].[O-]S([O-])(=O)=O |
Specifications
Blue | |
7778-18-9, 7646-79-9 | |
MFCD00010912 | |
15, 1708 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 | |
Drierite™, With indicator, 8 Mesh |
Granular | |
CaO4S | |
01,107 | |
SiO2 and insolubles: 0.58% | |
0.29% | |
calcium sulfate | |
24497 | |
136.13 |
Safety and Handling
GHS H Statement:
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage fertility.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effect
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Obtain special instructions before use.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only