Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
125.00 € - 204.00 €
Specifications
Material Cellulose Acetate
Type BuzzPlugs Bottle Closure
For Use With (Application) Minimizes evaporation, keeps food fresh longer
Includes Superior barrier
Quantity 1000 Case
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10346221
View Documents
Fisherbrand™
AS273
2.5cm narrow plastic shell vials
125.00 €
Pack of 1000
10550152
View Documents
Fisherbrand™
AS275
2.8cm wide plastic shell vials
204.00 €
Pack of 1000
Description
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
Specifications
Cellulose Acetate
Minimizes evaporation, keeps food fresh longer
1000 Case
BuzzPlugs Bottle Closure
Superior barrier
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures