Learn More
Oakton™ Economy Hygrothermograph
Product Code. 11708639 Shop All Cole-Parmer Products
missing translation for 'orderingAttributeHoverText'
:
Replacement red pen
Product Code. 11708639
missing translation for 'mfr': Oakton™ 0836880
Only null left
Add to basket
Add to basket
Use these replacement pens with Oakton™ Economy Hygrothermographs.
  • Red felt tip pens
  • Sold in packs of six
  • For Oakton™ Economy Hygrothermograph
Specifications
Includes 55 Sheets of seven-day, chart paper and one AA battery
For Use With (Application) Measuring and recording both humidity and temperature
Product Type Red Felt-tip Pen